missing translation for 'onlineSavingsMsg'
Learn More

Ethyl nipecotate, 97%

Product Code. 10277153 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10277153 500 g Glass bottle 500g
10700701 25 g Glass bottle 25g
10522021 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10277153 Supplier Thermo Scientific Acros Supplier No. 186625000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5006-62-2 | C8H15NO2 | 157.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5006-62-2
Molecular Formula C8H15NO2
Molecular Weight (g/mol) 157.21
MDL Number MFCD00005991
InChI Key XIWBSOUNZWSFKU-UHFFFAOYSA-N
Synonym ethyl nipecotate, ethyl 3-piperidinecarboxylate, nipecotic acid ethyl ester, 3-piperidinecarboxylic acid ethyl ester, ethylpiperidine-3-carboxylate, 3-ethoxycarbonyl piperidine, 3-piperidinecarboxylic acid, ethyl ester, piperidine-3-carboxylic acid ethyl ester, ethylnipecotate, a-ethyl nipecotate
PubChem CID 98969
IUPAC Name ethyl piperidine-3-carboxylate
SMILES CCOC(=O)C1CCCNC1

Specifications

Density 1.0100g/mL
Boiling Point 102°C to 104°C (7.0 mmHg)
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4580 to 1.461
Quantity 500 g
Specific Gravity 1.01
Solubility Information Solubility in water: miscible. Other solubilities: soluble in most organic solvents
Formula Weight 157.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethyl nipecotate
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl nipecotate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.