missing translation for 'onlineSavingsMsg'
Learn More

Ethyleneglycoldimethacrylate (98%, contains 90-110 ppm monomethyletherhydroquinone as inhibitor), TRC

Product Code. 30386671
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30386671 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30386671 Supplier Toronto Research Chemicals Supplier No. TRCE91752010ML
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyleneglycoldimethacrylate
CAS 97-90-5
Molecular Formula C10H14O4
Quantity 10 mL
Synonym 2-Propenoic acid, 2-methyl-, 1,1'-(1,2-ethanediyl) ester (ACI), 2-Propenoic acid, 2-methyl-, 1,2-ethanediyl ester (9CI), Methacrylic acid, ethylene ester (6CI, 8CI), 1 G, 1,2-Bis(methacryloyloxy)ethane, 1,2-Ethanediol dimethacrylate, 2-(2-Methylprop-2-enoyloxy)ethyl 2-methylprop-2-enoate, 2-[(2-Methylprop-2-enoyl)oxy]ethyl 2-methylprop-2-enoate, 2-Methacryloyloxyethyl methacrylate, Acryester ED, Acryester EDMA, Blemmer PDE 50, Diglycol dimethacrylate, DMM 2, EG 50, EGDMA, EM 320, Ethanediol dimethacrylate, Ethylene dimethacrylate, Ethylene glycol bis(methacrylate), Ethylene glycol dimethacrylate, Ethylene methacrylate, FA 121M, Fancryl FA 121M, Hi-Cross ED, Hi-Cross ED-P, Light Ester EG, Light Ester EG 50, Luvomaxx EDMA-DL 75, Miramer M 221, NK Ester 1G, NSC 24166, Perkalink 401, Perkalink 401-50D-pd, Perkalink 7401, San Ester EG, Saret 206, Sartomer SR 206, SR 206, SR 206NS, TGM 1, TGM 1 (ester)
InChI Formula InChI=1S/C10H14O4/c1-7(2)9(11)13-5-6-14-10(12)8(3)4/h1,3,5-6H2,2,4H3
SMILES CC(=C)C(=O)OCCOC(=O)C(=C)C
Molecular Weight (g/mol) 198.22
Formula Weight 198.0892
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.