missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic acid zinc disodium salt hydrate

Product Code. p-7049768
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11358047

Brand: Thermo Scientific Alfa Aesar 022426.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14025-21-9 | C10H12N2Na2O8Zn | 399.572 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14025-21-9
Molecular Formula C10H12N2Na2O8Zn
MDL Number MFCD00135187
InChI Key UBLOJEHIINPTTG-UHFFFAOYSA-J
Synonym zinc disodium edta, edta disodium zinc salt, unii-r84ov391ba, sequestrene na2zn zinc chelate, disodium zinc edetate, disodium zinc ethylenediaminetetraacetate, zinc disodium ethylenediaminetetraacetate, disodium zinc ethylenediaminetetraacetate dihydrate, acetic acid, ethylenedinitrilo tetra-, disodium salt, zinc complex, dihydrate, ethylenediaminetetraacetic acid disodium zinc salt
PubChem CID 51808
IUPAC Name disodium;zinc;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxylatomethyl)amino]acetate

Specifications

Quantity 25 g
SMILES [Na+].[Na+].[Zn++].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
Molecular Weight (g/mol) 399.57
Formula Weight 399.50 (Anhydrous)
Physical Form Powder
Chemical Name or Material Ethylenediaminetetraacetic acid zinc disodium salt hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.