missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic Acid Trisodium Salt Solution 0.1M (0.2N), NIST Standard Concentrate, for volumetric analysis, Fisher Chemical™

Product Code. 10121430 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 AMP
6 AMP
Unit Size:
1 ampoule
6 ampoules
This item is not returnable. View return policy

Product Code. 10121430

Brand: Fisher Chemical J/3700C/90

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Chemical Identifiers

CAS 150-38-9
Molecular Formula C10H19N2Na3O11
Molecular Weight (g/mol) 412.23
MDL Number MFCD00149685,MFCD02253006 (.3H2O)
InChI Key FXNQQEVEDZAAJM-UHFFFAOYSA-K
Synonym ethylenediaminetetraacetic acid trisodium salt, trisodium ethylene diamine tetraacetic acid
PubChem CID 131711402
IUPAC Name 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid;sodium
SMILES O.O.O.[Na+].[Na+].[Na+].OC(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O

Specifications

Color Colorless
pH 4 to 5
Physical Form Liquid
Quantity 6 AMP
Packaging Ampoule
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.