missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic acid trisodium salt, MedChemExpress

Product Code. 16462299
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16462299 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16462299 Supplier Medchem Express Supplier No. HYB1009

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ethylenediaminetetraacetic acid trisodium salt (EDTA trisodium salt) is used to bind metal ions in the practice of chelation therapy, for treating mercury and lead poisoning, used in a similar manner to remove excess iron from the body, for treating the complication of repeated blood transfusions, as would be applied to treat thalassaemia.

Ethylenediaminetetraacetic acid trisodium salt (EDTA trisodium salt) is used to bind metal ions in the practice of chelation therapy, for treating mercury and lead poisoning, used in a similar manner to remove excess iron from the body, for treating the complication of repeated blood transfusions, as would be applied to treat thalassaemia.

Specifications

Chemical Name or Material Ethylenediaminetetraacetic acid trisodium salt
Physical Form Solid
CAS 150-38-9
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C10H13N2Na3O8
Synonym EDTA trisodium salt Trisodium EDTA
Solubility Information H2O : 50 mg/mL (139.59 mM; Need ultrasonic)
SMILES O=C(O)CN(CCN(CC(O[Na])=O)CC(O[Na])=O)CC(O[Na])=O
Molecular Weight (g/mol) 358.19
Formula Weight 358.19
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.