missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ Ethylenediaminetetraacetic acid trisodium salt hydrate, 95%
Description
Applications
This is used to eliminate enzyme inhibition by traces of heavy metals, and to inhibit enzymes that require divalent cations as cofactors.
Solubility
Soluble in sodium chloride. (1 M NaOH).
Notes
Incompatible with oxidizing agents. Store at room temperature.
This is used to eliminate enzyme inhibition by traces of heavy metals, and to inhibit enzymes that require divalent cations as cofactors.
Solubility
Soluble in sodium chloride. (1 M NaOH).
Notes
Incompatible with oxidizing agents. Store at room temperature.
Specifications
Specifications
| Melting Point | >300°C |
| Quantity | 250 g |
| Formula Weight | 358.19 |
| Percent Purity | 95% |
| Chemical Name or Material | Ethylenediaminetetraacetic acid trisodium salt hydrate |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?