missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic acid tripotassium salt

Product Code. 11338047
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11338047 25 g 25g
11318047 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11338047 Supplier Thermo Scientific Alfa Aesar Supplier No. 022425.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 17572-97-3 | C10H13K3N2O8 | 406.52 g/mol

Ethylenediaminetetraacetic acid tripotassium salt is used to eliminate enzyme inhibition by traces of heavy metals, and to inhibit enzymes that require divalent cations as cofactors. And it is also used to study chelators, biochemicals and reagents. EDTA tripotassium salt has been used in a study to assess the safety of EDTA and HEDTA ingredients in cosmetic formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17572-97-3
Molecular Formula C10H13K3N2O8
Molecular Weight (g/mol) 406.52
MDL Number MFCD00013304
InChI Key FYZXEMANQYHCFX-UHFFFAOYSA-K
Synonym edetate tripotassium, tripotassium edta, edta tripotassium salt, ethylenediaminetetraacetic acid tripotassium salt, tripotassium hydrogen ethylenediaminetetraacetate, sequestrene 50k3, unii-kia5203u1h, tripotassium ethylenediaminetetraacetate, edta-3k, glycine, n,n'-1,2-ethanediylbis n-carboxymethyl-, tripotassium salt
PubChem CID 62717
IUPAC Name tripotassium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxymethyl)amino]acetate
SMILES [K+].[K+].[K+].OC(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O

Specifications

Odor Odorless
Quantity 25 g
Solubility Information Soluble in water,0.1 M at 20°C.
Formula Weight 406.46
Physical Form Crystalline
Chemical Name or Material Ethylenediaminetetraacetic acid tripotassium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.