missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ethylenediaminetetraacetic acid, tripotassium salt dihydrate, 99+%

Product Code. 10542461 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10542461 100 g Plastic bottle 100g
10552461 500 g Plastic bottle 500g
11985351 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10542461 Supplier Thermo Scientific Chemicals Supplier No. 301601000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 65501-24-8 | C10H13K3N2O8·2H2O | 442.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65501-24-8
Molecular Formula C10H13K3N2O8·2H2O
Molecular Weight (g/mol) 442.56
InChI Key MAPFUJCWRWFQIY-UHFFFAOYSA-K
Synonym ethylenediaminetetraacetic acid tripotassium salt dihydrate, edta tripotassium salt, edta tripotassium salt dihydrate, tripotassium ethylenediaminetetraacetate dihydrate, potassium 2,2'-2-carboxylatomethyl carboxymethyl amino ethyl azanediyl diacetate dihydrate, tripotassium 2-2-bis carboxylatomethyl amino ethyl carboxymethyl amino acetate dihydrate, edta tripotassium salt, dihydrate, ethylenediaminetetraacetic acid, tripotassium salt dihydrate, ethylenediaminetetraacetic acid tripotassium salt dihydrate titration
PubChem CID 2733162
IUPAC Name tripotassium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxymethyl)amino]acetate;dihydrate
SMILES C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)O)CC(=O)[O-].O.O.[K+].[K+].[K+]

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 99% min. (on dry substance) (Complexometry)
Packaging Plastic bottle
Linear Formula (KOCOCH2)2NCH2CH2N(CH2COOK)CH2COOH·2H2O
Quantity 100 g
Formula Weight 442.56
Percent Purity 99+%
Physical Form Fine Crystalline Powder
Chemical Name or Material Ethylenediaminetetraacetic acid, tripotassium salt dihydrate
compliance-icons
Product Identifier
  • Ethylenediaminetetraacetic acid, tripotassium salt dihydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Ethylenediaminetetraacetic acid, tripotassium salt dihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.