missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic acid tripotassium salt, Thermo Scientific Chemicals

Catalog Number p-7049767
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
17572-97-3
C10H13K3N2O8
406.52
MFCD00013304
FYZXEMANQYHCFX-UHFFFAOYSA-K
edetate tripotassium, tripotassium edta, edta tripotassium salt, ethylenediaminetetraacetic acid tripotassium salt, tripotassium hydrogen ethylenediaminetetraacetate, sequestrene 50k3, unii-kia5203u1h, tripotassium ethylenediaminetetraacetate, edta-3k, glycine, n,n'-1,2-ethanediylbis n-carboxymethyl-, tripotassium salt
62717
tripotassium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxymethyl)amino]acetate
[K+].[K+].[K+].OC(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
This item is not returnable. View return policy
17572-97-3
C10H13K3N2O8
406.52
MFCD00013304
FYZXEMANQYHCFX-UHFFFAOYSA-K
edetate tripotassium, tripotassium edta, edta tripotassium salt, ethylenediaminetetraacetic acid tripotassium salt, tripotassium hydrogen ethylenediaminetetraacetate, sequestrene 50k3, unii-kia5203u1h, tripotassium ethylenediaminetetraacetate, edta-3k, glycine, n,n'-1,2-ethanediylbis n-carboxymethyl-, tripotassium salt
62717
tripotassium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxymethyl)amino]acetate
[K+].[K+].[K+].OC(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O

Ethylenediaminetetraacetic acid tripotassium salt is used to eliminate enzyme inhibition by traces of heavy metals, and to inhibit enzymes that require divalent cations as cofactors. And it is also used to study chelators, biochemicals and reagents. EDTA tripotassium salt has been used in a study to assess the safety of EDTA and HEDTA ingredients in cosmetic formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Odor Odorless
Quantity 25 g
Solubility Information Soluble in water,0.1 M at 20°C.
Formula Weight 406.46
Physical Form Crystalline
Chemical Name or Material Ethylenediaminetetraacetic acid tripotassium salt

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'