missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
E4F1 Monoclonal specifically detects E4F1 in Human samples. It is validated for Flow Cytometry/Immunocytochemistry/Immunofluorescence/Protein Array.
Specifications
Specifications
| Antigen | E4F1 |
| Applications | Flow Cytometry, Immunocytochemistry/Immunofluorescence, Protein Array |
| Classification | Monoclonal |
| Clone | PCRP-E4F1-2D1 |
| Conjugate | PerCP |
| Formulation | PBS |
| Gene Alias | E4F transcription factor 1p50E4F, E4FMGC99614, EC 6.3.2.-, p120E4F, Putative E3 ubiquitin-protein ligase E4F1, Transcription factor E4F, transcription factor E4F1 |
| Host Species | Mouse |
| Immunogen | Recombinant full-length human E4F1 protein (Uniprot: Q66K89) |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?