missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Ethylenediaminetetraacetic acid, tetrasodium salt hydrate, 98%, pure
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Unit Size:
10kg
2.5kg
500g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
Specifications
Specifications
| Melting Point | >300.0°C |
| Color | White to Yellow |
| pH | 9.5 to 10.5 (1% soln.) |
| Physical Form | Crystalline Powder |
| Quantity | 2.5 kg |
| Assay Percent Range | 97.5% min. (on dry substance) (Complexometry) |
| Linear Formula | (NaO2CCH2)2NCH2CH2N(CH2CO2Na)2·xH2O |
| Fieser | 01,373; 05,296 |
| Merck Index | 14,3517 |
| Solubility Information | Solubility in water: 800g/L (20°C) |
| Show More |
Safety and Handling
Product Identifier
- Ethylenediaminetetraacetic acid, tetrasodium salt hydrate
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction