missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic Acid Tetrasodium Salt Dihydrate, TRC

Product Code. 30372717
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30372717 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30372717 Supplier Toronto Research Chemicals Supplier No. TRCE918310100G
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetrasodium Edetate Dihydrate
CAS 10378-23-1
Molecular Formula C10 H12 N2 O8 . 2 H2 O . 4 Na
Quantity 100 g
Synonym Glycine, N,N'-1,2-ethanediylbis[N-(carboxymethyl)-, sodium salt, hydrate (1:4:2) (ACI), Acetic acid, (ethylenedinitrilo)tetra-, tetrasodium salt, dihydrate (8CI), Glycine, N,N'-1,2-ethanediylbis[N-(carboxymethyl)-, tetrasodium salt, dihydrate (9CI), Tetrasodium ethylenediaminetetraacetate dihydrate, Tetrasodium Edetate Dihydrate, Ethylenediaminetetraacetic Acid Tetrasodium Salt Dihydrate
InChI Formula InChI=1S/C10H16N2O8.4Na.2H2O/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;;;;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;;;;2*1H2/q;4*+1;;/p-4
SMILES O.O.[Na+].[Na+].[Na+].[Na+].[O-]C(=O)CN(CCN(CC(=O)[O-])CC(=O)[O-])CC(=O)[O-]
IUPAC Name tetrasodium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate
Molecular Weight (g/mol) 416.2
Formula Weight 416.04
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.