missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic acid manganese disodium salt hydrate

Product Code. 11338508
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11338508 5 g 5g
11328508 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11338508 Supplier Thermo Scientific Alfa Aesar Supplier No. 040519.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Metal chelating compound | CAS: 15375-84-5 | C10H12MnN2Na2O8 | 389.13 g/mol

Ethylenediaminetetraacetic acid manganese disodium salt hydrate is a metal chelating compound. Cobalt was analysed complexometrically using ethylenediaminetetra-acetic acid (EDTA) by masking aluminum. Previous studies of manganese-EDTA and manganese-EGT-1 complexes indicated that the &-nitrogen coordination of EDTA resulted in a highly anisotropic spectrum for Mn(II). Lipophilic derivatives of manganese EDTA chelates when incorporated into liposomes confer the greatest hepatic enhancement per micromole of metal ion and have favorable clearance kinetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15375-84-5
Molecular Formula C10H12MnN2Na2O8
Molecular Weight (g/mol) 389.13
MDL Number MFCD00661156
InChI Key ZQHKAIDDHTYINE-UHFFFAOYSA-J
Synonym disodium manganese edta, edta disodium manganese salt, disodium manganese ethylenediaminetetraacetate, manganese disodium ethylene diamine tetraacetate, ethylenediaminetetraacetic acid, disodium manganese salt, disodium ethylenedinitrilo tetraacetato manganese, manganate 2-, disodium, glycine,n'-1,2-ethanediylbis n-carboxymethyl-, manganese disodium salt, manganate 2-,n'-1,2-ethanediylbis n-carboxymethyl glycinato 4--n,n',o,o',on,on'-, disodium, oc-6-21
PubChem CID 5148022
IUPAC Name 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid;manganese(2+)
SMILES [Na+].[Na+].[Mn++].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O

Specifications

Odor Odorless
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 389.13 (Anhydrous)
Physical Form Powder
Chemical Name or Material Ethylenediaminetetraacetic acid manganese disodium salt hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.