missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic acid, iron(III) monosodium salt

Product Code. 11399359
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
Unit Size:
100g
2kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11399359 2 kg 2kg
11379359 100 g 100g
11389359 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11399359 Supplier Thermo Scientific Alfa Aesar Supplier No. 088995.A3
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15708-41-5 | C10H12FeN2NaO8 | 367.047 g/mol

Ethylenediaminetetraacetic acid, iron(III) monosodium salt is used to eliminate enzyme inhibition by traces of heavy metals and to inhibit enzymes that require divalent cations as cofactors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15708-41-5
Molecular Formula C10H12FeN2NaO8
Molecular Weight (g/mol) 367.047
MDL Number MFCD00078215
InChI Key MKWYFZFMAMBPQK-UHFFFAOYSA-J
Synonym sodium feredetate, edta ferric sodium salt, calmosine, sodium ironedetate, edta iron iii sodium salt, ferisan, sytron, sequestrene nafe, sodium iron edta, ferric sodium edta
PubChem CID 27461
ChEBI CHEBI:78292
IUPAC Name sodium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;iron(3+)
SMILES C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)[O-])CC(=O)[O-].[Na+].[Fe+3]

Specifications

Odor Odorless
Quantity 2 kg
Solubility Information Soluble in water.
Formula Weight 367.05
Physical Form Powder
Chemical Name or Material Ethylenediaminetetraacetic acid, iron (III) monosodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.