missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ethylenediaminetetraacetic acid, ferric sodium salt trihydrate, 98%, pure

Product Code. 10175750 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Unit Size:
1kg
250g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10175750 250 g 250g
10287320 1 kg 1kg
10677554 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10175750 Supplier Thermo Scientific Chemicals Supplier No. 304682500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ethylenediaminetetraacetic acid, ferric sodium salt trihydrate, 98%, C10H12FeN2NaO8.3H2O, CAS Number-18154-32-, sodium ironedetate, edta ferric sodium salt, calmosine, sodium feredetate, edta iron iii sodium salt, sequestrene nafe, ferric sodium edta, ferisan, sodium iron edta, sytron, 1kg | CAS: 18154-32-0 | C10H12FeN2NaO8·3H2O | 421.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18154-32-0
Molecular Formula C10H12FeN2NaO8·3H2O
Molecular Weight (g/mol) 421.1
MDL Number MFCD00798117
InChI Key MKWYFZFMAMBPQK-UHFFFAOYSA-J
Synonym sodium feredetate, edta ferric sodium salt, calmosine, sodium ironedetate, edta iron iii sodium salt, ferisan, sytron, sequestrene nafe, sodium iron edta, ferric sodium edta
PubChem CID 27461
ChEBI CHEBI:78292
IUPAC Name sodium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;iron(3+)
SMILES C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)[O-])CC(=O)[O-].[Na+].[Fe+3]

Specifications

Color Brown-Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Quantity 250 g
Merck Index 15,4058
Solubility Information Solubility in water: soluble.
Formula Weight 421.1
Percent Purity 98%
Grade Pure
Physical Form Powder
Chemical Name or Material Ethylenediaminetetraacetic acid, ferric sodium salt trihydrate, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.