missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic Acid Disodium Salt Solution 0.01M (0.02N), NIST Standard Solution ready to use, for volumetric analysis, Fisher Chemical™

Catalog Number 10010190
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Bag in box
HDPE plastic bottle
139-33-3
C10H14N2Na2O8
336.21
MFCD00070672,MFCD00003541,MFCD00070672,MFCD00150037
ZGTMUACCHSMWAC-UHFFFAOYSA-L
ethylenediaminetetraacetic acid disodium salt, edta na2, disodium 2-2-bis carboxymethyl amino ethyl-carboxymethyl amino acetic acid
57339238
CHEBI:64734
disodium;2-[2-[bis(carboxylatomethyl)azaniumyl]ethyl-(carboxylatomethyl)azaniumyl]acetate
[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
139-33-3
C10H14N2Na2O8
336.21
MFCD00070672,MFCD00003541,MFCD00070672,MFCD00150037
ZGTMUACCHSMWAC-UHFFFAOYSA-L
ethylenediaminetetraacetic acid disodium salt, edta na2, disodium 2-2-bis carboxymethyl amino ethyl-carboxymethyl amino acetic acid
57339238
CHEBI:64734
disodium;2-[2-[bis(carboxylatomethyl)azaniumyl]ethyl-(carboxylatomethyl)azaniumyl]acetate
[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

Ethylenediaminetetraacetic Acid Disodium Salt Solution 0.01M (0.02N), NIST Standard Solution ready to use, for volumetric analysis

TRUSTED_SUSTAINABILITY
Color Colorless
Physical Form Liquid
Quantity 1 L
Packaging HDPE plastic bottle
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'