missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic Acid Disodium Salt Solution 0.01M (0.02N), NIST Standard Concentrate, for volumetric analysis, Fisher Chemical

Product Code. 10059981 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 Ampule
Packaging:
Ampoule
Unit Size:
1 ampoule
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10059981 1 Ampule Ampoule 1 ampoule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 10059981 Supplier Fisher Chemical Supplier No. J/3720C/05
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
For volumetric analysis
TRUSTED_SUSTAINABILITY

Specifications

CAS 139-33-3
Color Colorless
Packaging Ampoule
Molecular Formula C10H14N2Na2O8
MDL Number MFCD00070672,MFCD00003541,MFCD00070672,MFCD00150037
Quantity 1 Ampule
Synonym ethylenediaminetetraacetic acid disodium salt, edta na2, disodium 2-2-bis carboxymethyl amino ethyl-carboxymethyl amino acetic acid
InChI Key ZGTMUACCHSMWAC-UHFFFAOYSA-L
SMILES [Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
IUPAC Name disodium;2-[2-[bis(carboxylatomethyl)azaniumyl]ethyl-(carboxylatomethyl)azaniumyl]acetate
Molecular Weight (g/mol) 336.21
PubChem CID 57339238
ChEBI CHEBI:64734
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.