missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylenediaminetetraacetic Acid Disodium Salt Dihydrate, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Quantity | 50 g |
| UN Number | NONH for all modes of transport |
| Beilstein | 3900609 |
| Flash Point | >100°C (>212°F) |
| Solubility Information | Soluble in water (0.1M) |
| Formula Weight | 372.24g/mol |
| Grade | HPLC |
| Chemical Name or Material | Ethylenediaminetetraacetic Acid Disodium Salt Dihydrate |
Safety and Handling
Product Identifier
- EDTA disodium salt dihydrate
- Warning
- Acute toxicity Category 4
- Specific target organ toxicity after repeated exposure Category 2
- H332-Harmful if inhaled.
- H373-May cause damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : EDTA disodium salt dihydrate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction