missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic Acid, Disodium Salt Dihydrate, Certified AR for Analysis, Fisher Chemical™

Product Code. 10526383
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 
This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 

C10H18N2Na2O10, CAS Number-6381-92-6, 5kg, White, 372.23g/mol, 252 deg.C, 150037, Plastic keg, 4 to 6, Solid

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6381-92-6
Molecular Formula C10H18N2Na2O10
Molecular Weight (g/mol) 372.24
MDL Number 150037
InChI Key OVBJJZOQPCKUOR-UHFFFAOYSA-L
IUPAC Name disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
SMILES O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

Specifications

Melting Point 252°C
Color White
pH 4 to 6
Physical Form Solid
Quantity 1 kg
Formula Weight 372.23g/mol
Packaging Plastic powder jar
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'