missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic acid disodium salt, 0.200N (0.1M) Standardized solution

Product Code. 11364747
Change view
Click to view available options
Quantity:
1 L
4 L
Unit Size:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11364747 4 L 4 L
11354747 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11364747 Supplier Thermo Scientific Alfa Aesar Supplier No. 039783.K7
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 139-33-3 | C10H14N2Na2O8 | 336.21 g/mol

Ethylenediaminetetraacetic acid disodium salt is widely used in textile industry. Usually applied to dissolve limescale. It is applied in textile industry, pulp and paper industry and also in chelation therapy. In cosmetics, it acts as a sequestering agent. It acts as a corrosion inhibitor to carbon steel in the industries. It also acts as a food additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139-33-3
Molecular Formula C10H14N2Na2O8
Molecular Weight (g/mol) 336.21
MDL Number MFCD00070672,MFCD00003541,MFCD00070672,MFCD00150037
InChI Key ZGTMUACCHSMWAC-UHFFFAOYSA-L
Synonym ethylenediaminetetraacetic acid disodium salt, edta na2, disodium 2-2-bis carboxymethyl amino ethyl-carboxymethyl amino acetic acid
PubChem CID 57339238
ChEBI CHEBI:64734
IUPAC Name disodium;2-[2-[bis(carboxylatomethyl)azaniumyl]ethyl-(carboxylatomethyl)azaniumyl]acetate
SMILES [Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

Specifications

Color Colorless
Assay Percent Range 0.200N (0.1M)
Odor Odorless
Quantity 4 L
Solubility Information Miscible with water.
Formula Weight 336.22
Physical Form Liquid
Chemical Name or Material Ethylenediaminetetraacetic acid disodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.