missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic acid disodium magnesium salt hydrate

Product Code. 11362667
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11362667 100 g 100g
11352667 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11362667 Supplier Thermo Scientific Alfa Aesar Supplier No. L11857.22
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 194491-32-2 | C10H12MgN2Na2O8 | 358.50 g/mol

Ethylenediaminetetraacetic acid disodium magnesium salt hydrate is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals. It is used as chelating agents over a broad pH range in aqueous systems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethylenediaminetetraacetic acid disodium magnesium salt hydrate is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals. It is used as chelating agents over a broad pH range in aqueous systems.

Solubility
Soluble in water. Insoluble in acid and organic liquids

Notes
Incompatible with oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 194491-32-2
Molecular Formula C10H12MgN2Na2O8
Molecular Weight (g/mol) 358.50
MDL Number MFCD00078217
InChI Key AWNVVAMWLMUZOZ-UHFFFAOYSA-J
Synonym unii-ndt563s5vz, edta magnesium disodium, magnesium disodium edta, ndt563s5vz, magnesium disodium ethylenediaminetetraacetate, edta magnesium disodium salt hydrate, ethylenediaminetetraacetic acid magnesium disodium salt, ethylenediaminetetraacetic acid disodium magnesium salt, ethylenediaminetetraacetic acid disodium magnesium salt hydrate, magnesium sodium ethylenediaminetetraacetate
PubChem CID 161064
IUPAC Name magnesium;disodium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxylatomethyl)amino]acetate
SMILES [Na+].[Na+].[Mg++].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O

Specifications

Melting Point >300°C
Odor Odorless
Quantity 100 g
Solubility Information Soluble in water. Insoluble in acid and organic liquids
Formula Weight 358.50 (Anhydrous)
Chemical Name or Material Ethylenediaminetetraacetic acid disodium magnesium salt hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.