missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ethylenediaminetetraacetic acid, dipotassium salt dihydrate, 99%, extra pure

Product Code. 10175170 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10175170 500 g Plastic bottle 500g
10676792 100 g Plastic bottle 100g
10687424 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10175170 Supplier Thermo Scientific Chemicals Supplier No. 209745000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 25102-12-9 | C10H14K2N2O8·2H2O | 404.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25102-12-9
Molecular Formula C10H14K2N2O8·2H2O
Molecular Weight (g/mol) 404.46
MDL Number MFCD00150036
InChI Key JFROQFOCFOKDKU-UHFFFAOYSA-L
Synonym ethylenediaminetetraacetic acid dipotassium salt dihydrate, dipotassium dihydrogen ethylenediaminetetraacetate dihydrate, potassium 2,2'-2-bis carboxymethyl amino ethyl azanediyl diacetate dihydrate, edta dipotassium salt dihydrate, edta dipotassium salt, ethylenediaminetetraacetic acid,dipotassium salt dihydrate, potassium ethylenediaminetetraacetate dibasic, dipotassium ethylenediaminetetraacetate dihydrate, ethylenedinitrilo tetraacetic acid dipotassium salt, acmc-209ggx
PubChem CID 53486451
IUPAC Name dipotassium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate
SMILES C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)O)CC(=O)O.O.O.[K+].[K+]

Specifications

Melting Point 272.0°C
Color White
pH 4 to 5 (5% soln.)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Complexometry)
Packaging Plastic bottle
Linear Formula KO2CCH2N(CH2CO2H)CH2CH2N(CH2CO2K)CH2CO2H
Quantity 500 g
Merck Index 14, 3517
Solubility Information (10% in water) Clear colorless
Formula Weight 404.46
Percent Purity 99%
Grade Extra Pure
Physical Form Fine Crystalline Powder
Chemical Name or Material Ethylenediaminetetraacetic acid, dipotassium salt dihydrate, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Ethylenediaminetetraacetic acid, dipotassium salt dihydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.