missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic Acid Dianhydride, TRC

Product Code. 30381056
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381056 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30381056 Supplier Toronto Research Chemicals Supplier No. TRCE91759510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethylenediaminetetraacetic Acid Dianhydride
CAS 23911-25-3
Molecular Formula C10H12N2O6
Quantity 10 g
Synonym 4,4'-(1,2-Ethanediyl)bis-2,6-morpholinedione, 4,4'-Ethylenedi-2,6-morpholinedione, 1,2-Bis[2,6-dioxo-4-morpholinyl]ethane, 4,4'-Ethylenebis(2,6-dioxomorpholine), 4,4'-Ethylenebis(2,6-morpholinedione), EDTA Bisanhydride, EDTA Dianhydride, Ethylenediaminetetraacetic Dianhydride, N,N'-1,2-ethanediylbis[N-(carboxymethyl)-glycine Cyclic N,N:N',N'-Dianhydride
InChI Formula InChI=1S/C10H12N2O6/c13-7-3-11(4-8(14)17-7)1-2-12-5-9(15)18-10(16)6-12/h1-6H2
SMILES O=C(OC(C1)=O)CN1CCN2CC(OC(C2)=O)=O
IUPAC Name 4,4'-(ethane-1,2-diyl)bis(morpholine-2,6-dione)
Molecular Weight (g/mol) 256.21
Formula Weight 256.07
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.