missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic acid diammonium salt hydrate

Product Code. 11478146
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11478146 100 g 100g
11468146 25 g 25g
11488146 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11478146 Supplier GC Internal (Alfa Aesar) Supplier No. A15286.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 20824-56-0 | C10H24N4O9 | 344.321 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20824-56-0
Molecular Formula C10H24N4O9
Molecular Weight (g/mol) 344.321
MDL Number MFCD00150463
InChI Key WRSUMHYBOYWMTD-UHFFFAOYSA-N
Synonym ethylenediaminetetraacetic acid diammonium salt hydrate, edetic acid diamine hydrate, diazanium,2-2-bis carboxymethyl amino ethyl-carboxylatome, ammonium 2,2-2-bis carboxymethyl amino ethyl azanediyl diacetate hydrate
PubChem CID 71311375
IUPAC Name azane;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid;hydrate
SMILES C(CN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O.N.N.O

Specifications

Quantity 100 g
Beilstein 7785165
Formula Weight 326.31 (Anhydrous)
Chemical Name or Material Ethylenediaminetetraacetic acid diammonium salt hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.