missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic Acid Diammonium Salt, Extra Pure, SLR, Fisher Chemical

Product Code. 10315510 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10315510 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10315510 Supplier Fisher Chemical Supplier No. D/0500/48

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 215°C
CAS 20824-56-0
Color White
pH 4 to 5
Physical Form Powder/Solid
Quantity 100 g
Molecular Formula C10H22N4O8
MDL Number 36454
InChI Key KYQODXQIAJFKPH-UHFFFAOYSA-N
SMILES [NH4+].[NH4+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
IUPAC Name diammonium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate
Molecular Weight (g/mol) 326.31
Formula Weight 326.31g/mol
Packaging Plastic powder jar
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.