missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ethylenediaminetetraacetic acid, 99.5%, ACS reagent

Product Code. 10617201 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Plastic bottle
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10617201 1 kg Plastic bottle 1kg
10035750 5 kg Plastic bottle 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10617201 Supplier Thermo Scientific Chemicals Supplier No. 409930010
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 60-00-4 | C10H16N2O8 | 292.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-00-4
Molecular Formula C10H16N2O8
Molecular Weight (g/mol) 292.24
MDL Number MFCD00003541
InChI Key KCXVZYZYPLLWCC-UHFFFAOYSA-N
Synonym edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol
PubChem CID 6049
ChEBI CHEBI:42191
IUPAC Name 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid
SMILES OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

Specifications

Decomposition Information >150°C
Melting Point 220.0°C
Color White
Flash Point >100°C
Ignition Residue 0.2% max.
Infrared Spectrum Authentic
Assay Percent Range 99.50%
Packaging Plastic bottle
Linear Formula (HO2CCH2)2NCH2CH2N(CH2CO2H)2
Quantity 1 kg
Fieser 01,373
Merck Index 15, 3565
Solubility Information 0.005% max. Insoluble Matter (in dilute NH4OH)
Formula Weight 292.23
Vapor Pressure 0.013hPa at 20°C
Percent Purity 99.5%
Grade ACS Reagent
Physical Form Fine Crystalline Powder
Chemical Name or Material Ethylenediaminetetraacetic acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethylenediaminetetraacetic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H373-May cause damage to organs.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Ethylenediaminetetraacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.