missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic acid, 99%

Product Code. 11479562
Change view
Click to view available options
Quantity:
500 g
1000 g
5000 g
Unit Size:
1000g
5000g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11479562 1000 g 1000g
11408920 500 g 500g
11499562 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11479562 Supplier Thermo Scientific Alfa Aesar Supplier No. A10713.0B
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 60-00-4 | C10H16N2O8 | 292.24 g/mol

Ethylenediaminetetraacetic acid is used as a complexing agent for many cations. It prevents metal ion impurities and used to remove excess iron from the body. It acts as a sequestering agent in cosmetic products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethylenediaminetetraacetic acid is used as a complexing agent for many cations. It prevents metal ion impurities and used to remove excess iron from the body. It acts as a sequestering agent in cosmetic products.

Solubility
Soluble in sodium hydroxide, sodium carbonate, ammonia solution, acid and common organic solvents. Slightly soluble in water.

Notes
Stable. Incompatible with copper, copper alloys, nickel, aluminum, strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-00-4
Molecular Formula C10H16N2O8
Molecular Weight (g/mol) 292.24
MDL Number MFCD00003541
InChI Key KCXVZYZYPLLWCC-UHFFFAOYSA-N
Synonym edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol
PubChem CID 6049
ChEBI CHEBI:42191
IUPAC Name 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid
SMILES OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

Specifications

Melting Point ∼245°C (decomposition)
Linear Formula (HO2CCH2)2NCH2CH2N(CH2CO2H)2
Quantity 1000 g
Beilstein 1716295
Merck Index 14,3517
Solubility Information Soluble in sodium hydroxide,sodium carbonate,ammonia solution,acid and common organic solvents. Slightly soluble in water.
Formula Weight 292.24
Percent Purity 99%
Chemical Name or Material Ethylenediaminetetraacetic acid
compliance-icons compliance-icons
Product Identifier
  • Ethylenediaminetetraacetic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H373-May cause damage to organs.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Ethylenediaminetetraacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.