missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic Acid (0.5M Solution/pH 8.0), Fisher BioReagents

Product Code. 10073074
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
20 L
Packaging:
Nalgene Poly Bottle
Poly Bottle
Poly CUBE
Unit Size:
1 L
100mL
20 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10073074 20 L Poly CUBE 20 L
10306983 100 mL Poly Bottle 100mL
10182903 500 mL Poly Bottle 500mL
10628203 1 L Nalgene Poly Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10073074 Supplier Fisher BioReagents Supplier No. BP248220
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
EDTA is used extensively in molecular biology to minimize metal ion impurities in reaction buffers. Ideal for DNA work.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-00-4 , 7732-18-5
Molecular Formula C10H16N2O8
Molecular Weight (g/mol) 292.24
MDL Number MFCD00003541
InChI Key KCXVZYZYPLLWCC-UHFFFAOYSA-N
Synonym edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol
PubChem CID 6049
ChEBI CHEBI:42191
IUPAC Name 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid
SMILES OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

Specifications

Color Colorless
Linear Formula (HOOCCH2)2NCH2CH2N(CH2COOH)2
Concentration 0.475 to 0.525M
Merck Index 15, 3565
Formula Weight 292.25
Identification Pass Test
Packaging Poly CUBE
pH 8.0
Quantity 20 L
Physical Form Liquid
Chemical Name or Material Ethylenediamine Tetraacetic Acid, 0.5 M Solution, pH 8.0 (for DNA Work)
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethylenediaminetetraacetic Acid (0.5M Solution/pH 8.0)
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.