missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediamine Tetraacetic Acid, ACS, MP Biomedicals™

Product Code. 11482832 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11482832 500 g 500g
11472832 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11482832 Supplier MP Biomedicals Supplier No. 0215252190
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Ethylenediamine Tetraacetic Acid, ACS is an ACS grade buffer component and chelating agent.

  • Ethylenediamine tetraacetic acid (EDTA) is a calcium chelator used to eliminate inhibition of enzyme catalyzed reactions due to traces of heavy metals.
  • For use as an anticoagulant, disodium or tripotassium salts of EDTA are most commonly used.
  • EDTA prevents platelet aggregation and is therefore the preferred anticoagulant for platelet counts.
  • EDTA is an inhibitor of metalloproteases and metal-activated proteases at an effective concentration of 1 to 10 μM.
  • It acts as a chelator of the zinc ion in the active site of metalloproteases, but EDTA can also inhibit other metal ion-dependent proteases such as calcium-dependent cysteine proteases.
  • EDTA interferes with biological processes which are metal-dependent.
  • It is a buffer component.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-00-4
Molecular Formula C10H16N2O8
Molecular Weight (g/mol) 292.24
MDL Number MFCD00003541
InChI Key KCXVZYZYPLLWCC-UHFFFAOYSA-N
Synonym edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol
PubChem CID 6049
ChEBI CHEBI:42191
IUPAC Name 2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid
SMILES OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

Specifications

Color White
Density 650 kg/m3
Flash Point >100°C
Quantity 500 g
Solubility Information Soluble in water. 1.0X10+6 mg/L (miscible) at 25°C
Formula Weight 292.2
Vapor Pressure 7.57X10-17 mm Hg at 25°C
Percent Purity ≥99.4%
Grade ACS Grade
Physical Form Powder
For Use With (Application) Chelating agent, Buffer Component
Chemical Name or Material Ethylenediamine Tetraacetic Acid
Show More Show Less

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.