missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediamine P-Toluenesulfonate, TRC

Product Code. 30387192
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387192 2.5 g 2.5g
30370832 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30387192 Supplier Toronto Research Chemicals Supplier No. TRCE9178582.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14034-59-4
Molecular Formula C7 H8 O3 S . C2 H8 N2
Molecular Weight (g/mol) 232.2999
Synonym Ethane-1,2-diamine mono(4-methylbenzenesulphonate), 1,2-Ethanediamine, 4-methylbenzenesulfonate (1:1), 1,2-Ethanediamine, mono(4-methylbenzenesulfonate) (9CI), Ethylenediamine, mono-p-toluenesulfonate (8CI), 2-Aminoethylammonium p-toluenesulfonate, Ethylenediamine monotosylate, Ethylenediamine p-toluenesulfonate, NSC 14086
IUPAC Name ethane-1,2-diamine;4-methylbenzenesulfonic acid
SMILES Cc1ccc(cc1)S(=O)(=O)O.NCCN

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C7H8O3S.C2H8N2/c1-6-2-4-7(5-3-6)11(8,9)10;3-1-2-4/h2-5H,1H3,(H,8,9,10);1-4H2
Formula Weight 232.0882
Chemical Name or Material Ethane-1,2-diamine Mono(4-methylbenzenesulphonate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.