missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylenediamine-N,N,N’,N’-tetraacetic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | Edetic Acid |
| CAS | 60-00-4 |
| Molecular Formula | C10 H16 N2 O8 |
| Quantity | 25 g |
| Synonym | Glycine, N,N'-1,2-ethanediylbis[N-(carboxymethyl)-, Acetic acid, (ethylenedinitrilo)tetra- (8CI), N,N'-1,2-Ethanediylbis[N-(carboxymethyl)glycine], 2,2',2'',2'''-(ethane-1,2-diylbis(azanetriyl))tetraacetic acid, 3,6-Diazaoctanedioic acid, 3,6-bis(carboxymethyl)-, Acetic acid, 2,2',2'',2'''-(1,2-ethanediyldinitrilo)tetrakis-, Acroma DH 700, Celon A, Celon ATH, Cheelox, Chelest 3A, Chelest A, Chemcolox 340, Clewat TAA, Clewat TTA, Complexon II, Dissolvine E, Dissolvine NA-X, Dissolvine Z, Dotite 4H, EDETA B, EDTA, EDTA (chelating agent), Edathamil, Edetic acid, Endrate, Ethylene-N,N'-biscarboxymethyl-N,N'-diglycine, Ethylenediamine-N,N,N',N'-tetraacetic acid, Ethylenediaminetetraacetate, Ethylenediaminetetraacetic acid, Ethylenedinitrilotetraacetic acid, Gluma Cleanser, Havidote, ICRF 185, Metaquest A, NSC 97243, NSC 97404, Nervanaid B acid, Nullapon B acid, Nullapon BF acid, Perma Kleer 50 acid, PrefGel, Quastal Special, Sequestrene AA, Sequestrene K 4, Sequestric acid, Sequestrol, Techrun DO, Titriplex, Titriplex II, Trilon BS, Trilon BW, Versene, WS, WS (chelating agent), YD 30, Zonon AO |
| InChI Formula | InChI=1S/C10H16N2O8/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20) |
| SMILES | OC(=O)CN(CCN(CC(=O)O)CC(=O)O)CC(=O)O |
| IUPAC Name | 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid |
| Molecular Weight (g/mol) | 292.24 |
| Formula Weight | 292.0907 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?