missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediamine dihydrochloride, 98%

Product Code. 11440064
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11440064 100 g 100g
11450064 500 g 500g
11460064 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11440064 Supplier Thermo Scientific Alfa Aesar Supplier No. A13563.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 333-18-6 | C2H10Cl2N2 | 133.02 g/mol

Ethylenediamine dihydrochloride was used in modified ethylenediamine condensation method for fluorimetric determination of catecholamines. It was used to investigate the luminescence properties of complexes of EuIII and TbIII with ethylenediamine. It was used in the synthesis of 1,3,5-tris(4,5-dihydro-1H-imidazol-2-yl)benzene3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethylenediamine dihydrochloride was used in modified ethylenediamine condensation method for fluorimetric determination of catecholamines. It was used to investigate the luminescence properties of complexes of EuIII and TbIII with ethylenediamine. It was used in the synthesis of 1,3,5-tris(4,5-dihydro-1H-imidazol-2-yl)benzene3

Solubility
Soluble in water 300 g/L (20°C).

Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 333-18-6
Molecular Formula C2H10Cl2N2
Molecular Weight (g/mol) 133.02
MDL Number MFCD00012524
InChI Key OHHBFEVZJLBKEH-UHFFFAOYSA-N
Synonym ethylenediamine dihydrochloride, chlor-ethamine, ethylenediammonium chloride, ethylenediammonium dichloride, ethane-1,2-diamine dihydrochloride, dimethylenediamine dihydrochloride, 1,2-diaminoethane dihydrochloride, 1,2-ethanediamine, dihydrochloride, ethylenediamine 2hcl, dimethylenediamine diydrochloride
PubChem CID 9521
ChEBI CHEBI:53626
IUPAC Name ethane-1,2-diamine;dihydrochloride
SMILES [H+].[H+].[Cl-].[Cl-].NCCN

Specifications

Melting Point >300°C
Quantity 100 g
Sensitivity Hygroscopic
Solubility Information Soluble in water 300g/L (20°C).
Formula Weight 133.02
Percent Purity 98%
Chemical Name or Material Ethylenediamine dihydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.