missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediamine dihydrochloride, 98+%

Product Code. 10357510 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10357510 100 g Plastic bottle 100g
10497560 500 g Plastic bottle 500g
10462845 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10357510 Supplier Thermo Scientific Acros Supplier No. 187651000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 333-18-6 | C2H10Cl2N2 | 133.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 333-18-6
Molecular Formula C2H10Cl2N2
Molecular Weight (g/mol) 133.02
MDL Number MFCD00012524
InChI Key OHHBFEVZJLBKEH-UHFFFAOYSA-N
Synonym ethylenediamine dihydrochloride, chlor-ethamine, ethylenediammonium chloride, ethylenediammonium dichloride, ethane-1,2-diamine dihydrochloride, dimethylenediamine dihydrochloride, 1,2-diaminoethane dihydrochloride, 1,2-ethanediamine, dihydrochloride, ethylenediamine 2hcl, dimethylenediamine diydrochloride
PubChem CID 9521
ChEBI CHEBI:53626
SMILES [H+].[H+].[Cl-].[Cl-].NCCN

Specifications

Melting Point >300.0°C
Color White
pH 3.9 to 5 (1% aq. soln.)
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Plastic bottle
Linear Formula H2NCH2CH2NH2·2HCl
Quantity 100 g
Beilstein 04, 230
Merck Index 15, 3849
Solubility Information (1% in water) Clear colorless
IUPAC Name ethane-1,2-diamine;dihydrochloride
Formula Weight 133.04
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material Ethylenediamine dihydrochloride, 98+%
Show More Show Less
compliance-icons
Product Identifier
  • Ethylenediamine dihydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.