missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediamine dihydrochloride, 98+%

Product Code. p-129171 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10357510

Brand: Thermo Scientific Acros 187651000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 333-18-6 | C2H10Cl2N2 | 133.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 333-18-6
Molecular Formula C2H10Cl2N2
Molecular Weight (g/mol) 133.02
MDL Number MFCD00012524
InChI Key OHHBFEVZJLBKEH-UHFFFAOYSA-N
Synonym ethylenediamine dihydrochloride, chlor-ethamine, ethylenediammonium chloride, ethylenediammonium dichloride, ethane-1,2-diamine dihydrochloride, dimethylenediamine dihydrochloride, 1,2-diaminoethane dihydrochloride, 1,2-ethanediamine, dihydrochloride, ethylenediamine 2hcl, dimethylenediamine diydrochloride
PubChem CID 9521
ChEBI CHEBI:53626
SMILES [H+].[H+].[Cl-].[Cl-].NCCN

Specifications

Melting Point >300.0°C
Color White
pH 3.9 to 5 (1% aq. soln.)
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Plastic bottle
Linear Formula H2NCH2CH2NH2·2HCl
Quantity 100 g
Beilstein 04, 230
Merck Index 15, 3849
Solubility Information (1% in water) Clear colorless
IUPAC Name ethane-1,2-diamine;dihydrochloride
Formula Weight 133.04
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material Ethylenediamine dihydrochloride, 98+%
Show More Show Less
compliance-icons
Product Identifier
  • Ethylenediamine dihydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.