missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylene Terephthalate Cyclic Trimer-d12, TRC
Specifications
Specifications
| Chemical Name or Material | Ethylene Terephthalate Cyclic Trimer-d12 |
| Molecular Formula | C30H12D12O12 |
| Quantity | 10 mg |
| Synonym | 3,6,13,16,23,26-Hexaoxatetracyclo[26.2.2.28,11.218,21]hexatriaconta-8,10,18,20,28,30,31,33,35-nonaene-2,7,12,17,22,27-hexone-d12, Terephthalic acid, trimol. cyclic ethylene ester, Ethylene glycol, trimol. cyclic terephthalate-d12, 1,4,11,14,21,24-Hexaoxatribenzo[abc,klm,uvw]cyclotriacontane-5,10,15,20,25,30-hexone, Cyclic ethylene terephthalate trimer-d12, Cyclic tris(ethylene terephthalate)-d12, Cyclotris(ethylene glycol terephthalate)-d12, Ethylene glycol-terephthalic acid cyclic trimer-d12, Ethylene terephthalate cyclic trimer-d12, Ethylene terephthalate trimer-d12, Tris ethylene terephthalate-d12 |
| InChI Formula | InChI=1S/C30H24O12/c31-25-19-1-2-20(4-3-19)26(32)38-15-16-40-28(34)22-9-11-24(12-10-22)30(36)42-18-17-41-29(35)23-7-5-21(6-8-23)27(33)39-14-13-37-25/h1-12H,13-18H2/i13D2,14D2,15D2,16D2,17D2,18D2 |
| SMILES | O=C(OC([2H])([2H])C([2H])([2H])OC(C1=CC=C(C(OC([2H])([2H])C([2H])([2H])OC(C2=CC=C3C=C2)=O)=O)C=C1)=O)C4=CC=C(C(OC([2H])([2H])C([2H])([2H])OC3=O)=O)C=C4 |
| Molecular Weight (g/mol) | 588.58 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?