missing translation for 'onlineSavingsMsg'
Learn More

Ethylene glycol, Spectrophotometric grade, 99+%, Thermo Scientific Chemicals

Catalog Number 11495270
Click to view available options
Quantity:
1 L
4 x 1 L
This item is not returnable. View return policy
107-21-1
C2H6O2
62.068
MFCD00002885
LYCAIKOWRPUZTN-UHFFFAOYSA-N
ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
174
CHEBI:30742
ethane-1,2-diol
C(CO)O
This item is not returnable. View return policy
107-21-1
C2H6O2
62.068
MFCD00002885
LYCAIKOWRPUZTN-UHFFFAOYSA-N
ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
174
CHEBI:30742
ethane-1,2-diol
C(CO)O

For use in spectrophotometryEthylene glycol is used in spectrophotometry and environmental testing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point °C to 13°C
Density 1.113 g/mL
Boiling Point 196°C to 198°C
Flash Point 119°C (246°F)
Assay Percent Range >99%
Refractive Index 1.431
Quantity 4 x 1 L
Beilstein 505945
Sensitivity Hygroscopic
Solubility Information Miscible with water,toluene,tetrahydrofuran,alcohol,chloroform and other organic solvents.
Formula Weight 62.07
Grade Spectrophotometric
Physical Form Liquid
Chemical Name or Material Ethylene glycol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'