missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Glycol, puriss. p.a., Reag. Ph. Eur., ≥99.5%, Honeywell™ Riedel-de Haen™
BIOPHARMA

Product Code. p-7120431
Click to view available options
Quantity:
250 mL
1 L
2.5 L
5 L
6 x 1 L
Packaging:
Poly Bottle
Unit Size:
1 L
2.5 L
250mL
5 L
Pack of 6
This item is not returnable. View return policy

Product Code. 15660690

Brand: Honeywell-Riedel-de Haen 03750250ML

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss. p.a., Reag. Ph. Eur.,≥99.5%

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-21-1
Molecular Formula C2H6O2
Molecular Weight (g/mol) 62.068
MDL Number MFCD00002885
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
PubChem CID 174
ChEBI CHEBI:30742
IUPAC Name ethane-1,2-diol
SMILES C(CO)O

Specifications

pH Neutral
Melting Point -12°C
Color Colorless
Density 1.113g/mL at 25°C
Boiling Point 197°C
Quantity 250 mL
Explosion Limit 0.153
Flash Point 111°C (231.8°F)
Ignition Point 752°F
Packaging Poly Bottle
Physical Form Liquid
Refractive Index n20/D 1.431
Linear Formula HOCH2CH2OH
Vapor Pressure 0.08mmHg (20°C)
UN Number UN3082
Beilstein 505945
Formula Weight 62.07g/mol
Percent Purity ≥99.5%
Grade puriss. p.a.
Chemical Name or Material Ethylene Glycol, Reag. Ph. Eur.
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : ethylene glycol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.