missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Glycol Monostearate, TRC

Product Code. 30137555
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137555 250 mg 250mg
30133343 100 mg 100mg
17955075 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137555 Supplier Toronto Research Chemicals Supplier No. TRCE917480250MG
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-60-4
Molecular Formula C20 H40 O3
Molecular Weight (g/mol) 328.53
Synonym Stearic acid, 2-hydroxyethyl ester (6CI,7CI,8CI), Ethylene glycol, monostearate (8CI), 2-Hydroxyethyl stearate, Cutina EGMS, EGMS 70V, Emalex EG-M-S, Emcol ES 50, Emerest 2350, Empilan 2848, Ethylene glycol stearate, Glycol monostearate, Glycol stearate, Ivorit, Kessco EGMS, Kessco EGMS 70, Monthybase, Monthyle, Nikkol EGMS 70, Parastarin, Prodhybas N, Prodhybase ethyl, S 151, Sedetol, Stepan EGMS, Tegin EGMS-VA, Tego-stearate, Octadecanoic acid, 2-hydroxyethyl ester
IUPAC Name 2-hydroxyethyl octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OCCO

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C20H40O3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-20(22)23-19-18-21/h21H,2-19H2,1H3
Formula Weight 328.2977
Chemical Name or Material Ethylene Glycol Monostearate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.