missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Glycol Diethyl Ether, 98%, Honeywell™

Product Code. 15692230 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15692230 500 mL 500mL
15682230 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15692230 Supplier Honeywell Research Chemicals Supplier No. 224111500ML
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 629-14-1
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.176
MDL Number MFCD00009253
InChI Key LZDKZFUFMNSQCJ-UHFFFAOYSA-N
Synonym ethylene glycol diethyl ether, diethyl cellosolve, ethane, 1,2-diethoxy, hisolve eme, 3,6-dioxaoctane, glyme-1, diethyl glycol, 2-ethoxyethyl ethyl ether, egdee, diethylether ethylenglykolu
PubChem CID 12375
IUPAC Name 1,2-diethoxyethane
SMILES CCOCCOCC

Specifications

Color Yellow
Density 0.842g/mL at 25°C
Boiling Point 121°C
Flash Point 22°C (71.6°F)
Packaging Glass Bottle
Linear Formula CH3CH2OCH2CH2OCH2CH3
Quantity 500 mL
UN Number UN1153
Beilstein 1732917
Formula Weight 118.17g/mol
Vapor Pressure 9.4mmHg (20°C)
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethylene Glycol Diethyl Ether
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.