missing translation for 'onlineSavingsMsg'
Learn More

Ethylene glycol-bis(beta-aminoethyl ether)-N,N,N',N'-tetraacetic acid tetrasodium salt, 95%

Product Code. 15720879 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15720879 50 g Glass bottle 50g
10517800 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15720879 Supplier Thermo Scientific Acros Supplier No. 428570500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13368-13-3 | C14H20N2Na4O10 | 468.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13368-13-3
Molecular Formula C14H20N2Na4O10
Molecular Weight (g/mol) 468.28
MDL Number MFCD00274420
InChI Key BAOGCDPNNOBWQZ-UHFFFAOYSA-J
Synonym egta tetrasodium, 3,12-bis carboxymethyl-6,9-dioxa-3,12-diazatetradecane-1,14-dioic acid, tetrasodium salt, ethylene glycol-bis beta-aminoethyl ether-n,n,n',n'-tetraacetic acid tetrasodium salt, egta tetrasodium salt, egtazic acid tetrasodium, ethylene-bis oxyethylenenitrilo tetraacetic acid tetrasodium, 2,2',2,2'-ethylenebis oxyethylenenitrilo tetraacetic acid tetrasodium salt, 6,9-dioxa-3,12-diazatetradecanedioicacid,3,12-bis carboxymethyl-,sodium salt 1:4, tetrasodium 3,12-bis carboxylatomethyl-6,9-dioxa-3,12-diazatetradecane-1,14-dioate, ethylene glycol-bis, a-aminoethyl ether-n,n,n inverted exclamation marka,n inverted exclamation marka-tetraacetic acid tetrasodium salt
PubChem CID 16219327
IUPAC Name tetrasodium;2-[2-[2-[2-[bis(carboxylatomethyl)amino]ethoxy]ethoxy]ethyl-(carboxylatomethyl)amino]acetate
SMILES C(COCCOCCN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)[O-])CC(=O)[O-].[Na+].[Na+].[Na+].[Na+]

Specifications

Melting Point 54.0°C to 56.0°C
Assay Percent Range 18.46 to 20.82% (ICP on dried sample)
Packaging Glass bottle
Quantity 50 g
Solubility Information Solubility in water: soluble.
Formula Weight 468.28
Percent Purity 95%
Chemical Name or Material Ethylene glycol-bis(β-aminoethyl ether)-N,N,N',N'-tetraacetic acid tetrasodium salt
compliance-icons
Product Identifier
  • Ethylene glycol-bis(beta-aminoethyl ether)-N,N,N',N'-tetraacetic acid tetrasodium salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Ethylene glycol-bis(beta-aminoethyl ether)-N,N,N',N'-tetraacetic acid tetrasodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.