missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Glycol-bis-(β-Aminoethylether)-N,N,N',N'- tetraacetic acid, Molecular Biology Reagent Grade, MP Biomedicals™

Product Code. 11446541 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
Unit Size:
100g
10g
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11446541 10 g 10g
11456541 25 g 25g
11466541 100 g 100g
11476541 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11446541 Supplier MP Biomedicals Supplier No. 0219482310
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 67-42-5
Molecular Formula C14H24N2O10
Molecular Weight (g/mol) 380.35
MDL Number MFCD00004291
InChI Key DEFVIWRASFVYLL-UHFFFAOYSA-N
Synonym egta, egtazic acid, gedta, ethylenebis oxyethylenenitrilo tetraacetic acid, ebonta, 6,9-dioxa-3,12-diazatetradecanedioic acid, 3,12-bis carboxymethyl, 1,2-bis 2-bis carboxymethyl amino ethoxy ethane, ethylene glycol tetraacetic acid, h4egta, egtazic acid usan:inn
PubChem CID 6207
ChEBI CHEBI:30740
IUPAC Name 2-[2-[2-[2-[bis(carboxymethyl)amino]ethoxy]ethoxy]ethyl-(carboxymethyl)amino]acetic acid
SMILES C(COCCOCCN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O

Specifications

Color White
Quantity 10 g
Formula Weight 380.4
Grade Molecular Biology Reagent
Physical Form Powder
Chemical Name or Material Ethylene Glycol-bis-(β-Aminoethylether)-N,N,N',N'- tetraacetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.