missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Glycol, ≥99%, Spectrophotometric Grade, Honeywell Riedel-de Haën™

Product Code. 15692950
Change view
Click to view available options
Quantity:
1 L
2 L
4 x 2 L
6 x 1 L
Packaging:
Glass Bottle
Unit Size:
1 L
2 L
Pack of 4
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15692950 2 L Glass Bottle 2 L
15682950 1 L Glass Bottle 1 L
15612960 6 x 1 L Glass Bottle Pack of 6
15602960 4 x 2 L Glass Bottle Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15692950 Supplier Honeywell Riedel-de Haën Supplier No. 2932372L
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Spectrophotometric Grade,≥99%

  • Spectrophotometric grade solvents are high purity solvents made specifically for applications using spectrophotometers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-21-1
Molecular Formula C2H6O2
Molecular Weight (g/mol) 62.068
MDL Number MFCD00002885
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
PubChem CID 174
ChEBI CHEBI:30742
IUPAC Name ethane-1,2-diol
SMILES C(CO)O

Specifications

Melting Point -12°C
Density 1.113g/mL at 25°C
Quantity 2 L
Explosion Limit 0.153
Flash Point 111°C (231.8°F)
Ignition Point 752°F
Packaging Glass Bottle
Linear Formula HOCH2CH2OH
Vapor Pressure 0.08mmHg (20°C)
UN Number UN3082
Beilstein 505945
Boiling Point 195°C to 198°C
Formula Weight 62.07g/mol
Percent Purity ≥99%
Grade Spectrophotometry
Chemical Name or Material Ethylene Glycol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.