missing translation for 'onlineSavingsMsg'
Learn More

Ethylene glycol, Reagent Grade, ≥99%, Honeywell™

Product Code. 15620960 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
18 L
Unit Size:
18 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15620960 18 L 18 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15620960 Supplier Honeywell Research Chemicals Supplier No. 10246618LCS

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 107-21-1
Density 1.113g/mL at 25°C
Quantity 18 L
Explosion Limit 0.153
Flash Point 111°C (231.8°F)
Ignition Point 752°F
Molecular Formula C2H6O2
Linear Formula HOCH2CH2OH
Vapor Pressure 0.08mmHg (20°C)
MDL Number MFCD00002885
UN Number UN3082
Beilstein 505945
Synonym ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILES C(CO)O
IUPAC Name ethane-1,2-diol
Molecular Weight (g/mol) 62.068
PubChem CID 174
ChEBI CHEBI:30742
Formula Weight 62.07g/mol
Percent Purity ≥99%
Grade ReagentPlus
Chemical Name or Material Ethylene Glycol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.