missing translation for 'onlineSavingsMsg'
Learn More

Ethylene glycol, 99% min., MP Biomedicals™

Product Code. 11426882 Shop All MP Biomedicals Products
Click to view available options
Quantity:
250 mL
1 L
Unit Size:
1 L
250mL

Product Code. 11426882

Brand: MP Biomedicals 0215108983

Please to purchase this item. Need a web account? Register with us today!

Chemical Identifiers

CAS 107-21-1
Molecular Formula C2H6O2
Molecular Weight (g/mol) 62.068
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
PubChem CID 174
ChEBI CHEBI:30742
IUPAC Name ethane-1,2-diol
SMILES C(CO)O

Specifications

Color Colorless
Density 1.113g/cm3 at 20°C
Quantity 250 mL
Physical Form Liquid
Assay Percent Range 99% min.
Formula Weight 62.07
Chemical Name or Material Ethylene glycol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.