missing translation for 'onlineSavingsMsg'
Learn More

Ethylene glycol, 99%

Product Code. 11428920
Change view
Click to view available options
Quantity:
10,000 g
250 g
2500 g
500 g
Unit Size:
10000g
2500g
250g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11428920 250 g 250g
11464663 500 g 500g
11474663 2500 g 2500g
11484663 10,000 g 10000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11428920 Supplier Thermo Scientific Alfa Aesar Supplier No. A11591.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 107-21-1 | C2H6O2 | 62.068 g/mol

Ethylene glycol is used as a precursor for the syntheses of polymers like polyester fibers, resins and polyethylene terephthalate, which finds application in making plastic bottles. It acts as an intermediate in the synthesis of 1,4-dioxane. It plays an important role in organic synthesis as a protecting group for carbonyl groups. It is a useful desiccant due to its high boiling point and affinity towards water. It plays an important role to remove water vapor from natural gas before proceeding for further processes. Its major application is as a medium for convective heat transfer in automobiles and liquid cooled computers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethylene glycol is used as a precursor for the syntheses of polymers like polyester fibers, resins and polyethylene terephthalate, which finds application in making plastic bottles. It acts as an intermediate in the synthesis of 1,4-dioxane. It plays an important role in organic synthesis as a protecting group for carbonyl groups. It is a useful desiccant due to its high boiling point and affinity towards water. It plays an important role to remove water vapor from natural gas before proceeding for further processes. Its major application is as a medium for convective heat transfer in automobiles and liquid cooled computers.

Solubility
Miscible with water, toluene, tetrahydrofuran, alcohol, chloroform and other organic solvents.

Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong acids, strong oxidizing agents, strong bases, aldehydes and aluminum.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-21-1
Molecular Formula C2H6O2
Molecular Weight (g/mol) 62.068
MDL Number MFCD00002885
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
PubChem CID 174
ChEBI CHEBI:30742
IUPAC Name ethane-1,2-diol
SMILES C(CO)O

Specifications

Melting Point -13°C
Density 1.113
Quantity 250 g
Flash Point 119°C (246°F)
Odor Sweet
Refractive Index 1.431
Linear Formula HOCH2CH2OH
Beilstein 505945
Merck Index 14,3798
Boiling Point 196°C to 198°C
Solubility Information Miscible with water,toluene,tetrahydrofuran,alcohol,chloroform and other organic solvents.
Formula Weight 62.07
Percent Purity 99%
Chemical Name or Material Ethylene glycol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethylene glycol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H373-May cause damage to organs.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Supplemental information
  • MIXTURE LIST-Contain: ethylene glycol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.