missing translation for 'onlineSavingsMsg'
Learn More

Ethylene dimethacrylate, 98%, stabilized

Product Code. 10177323 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
5 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
250mL
5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10177323 1 L Glass bottle 1 L
10607201 250 mL Glass bottle 250mL
13418558 5 L Plastic drum 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10177323 Supplier Thermo Scientific Acros Supplier No. 409920010
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 97-90-5 | C10H14O4 | 198.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-90-5
Molecular Formula C10H14O4
Molecular Weight (g/mol) 198.22
MDL Number MFCD00008590
InChI Key STVZJERGLQHEKB-UHFFFAOYSA-N
Synonym ethylene glycol dimethacrylate, ethylene dimethacrylate, glycol dimethacrylate, diglycol dimethacrylate, ethanediol dimethacrylate, ethylenedimethyacrylate, ethylene methacrylate, ethyldiol metacrylate, 1,2-bis methacryloyloxy ethane, ethylene glycol bis methacrylate
PubChem CID 7355
ChEBI CHEBI:53436
SMILES CC(=C)C(=O)OCCOC(=O)C(C)=C

Specifications

Melting Point -40.0°C
Color Colorless to Yellow
Density 1.0500g/mL
Boiling Point 98.0°C to 100.0°C (5.0 mmHg)
Flash Point 101°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (H2C=C(CH3)COOCH2-)2
Refractive Index 1.4530 to 1.4550
Quantity 1 L
Specific Gravity 1.05
Solubility Information Solubility in water: soluble.
IUPAC Name 2-[(2-methylprop-2-enoyl)oxy]ethyl 2-methylprop-2-enoate
Stabilizer 90 to 110ppm MEHQ
Viscosity 3.1 mPa.s (20°C)
Formula Weight 198.22
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethylene dimethacrylate, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.