missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Diamine, TRC

Product Code. 30379676
Change view
Click to view available options
Quantity:
500 mL
1 L
Unit Size:
1 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30379676 500 mL 500mL
30385209 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30379676 Supplier Toronto Research Chemicals Supplier No. TRCE917695500ML
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethylenediamine
CAS 107-15-3
Purity Grade Notes GC
Molecular Formula C2 H8 N2
Quantity 500 mL
Synonym Ethylenediamine, Ethane-1,2-diamine, 1,2-Ethanediamine, Ethylenediamine (8CI), (2-Aminoethyl)amine, 1,2-Diaminoethane, 1,2-Ethylenediamine, 1,4-Diazabutane, Dimethylenediamine, EDA, Edamine, LR 500G, PB 05721, α,ω-Ethanediamine, β-Aminoethylamine
InChI Formula InChI=1S/C2H8N2/c3-1-2-4/h1-4H2
SMILES NCCN
IUPAC Name ethane-1,2-diamine
Molecular Weight (g/mol) 60.1
Formula Weight 60.0687
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.