missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylene-d4 Carbonate, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 1,3-Dioxolan-2-one-d4 |
| CAS | 362049-63-6 |
| Molecular Formula | C3 2H4 O3 |
| Quantity | 0.25 g |
| InChI Formula | InChI=1 S/C3H4O3/c4-3-5-1-2-6-3/h1-2H2/i1D2,2D2 |
| SMILES | [2 H]C1([2 H])OC(=O)OC1([2 H])[2 H] |
| IUPAC Name | 4,4,5,5-tetradeuterio-1,3-dioxolan-2-one |
| Molecular Weight (g/mol) | 92.09 |
| Formula Weight | 92.0412 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction