Learn More
Ethyl N-piperazinecarboxylate, 99%, Thermo Scientific Chemicals
£28.00 - £202.00
Chemical Identifiers
CAS | 120-43-4 |
---|---|
Molecular Formula | C7H14N2O2 |
Molecular Weight (g/mol) | 158.2 |
MDL Number | MFCD00005964 |
InChI Key | LNOQURRKNJKKBU-UHFFFAOYSA-N |
Synonym | ethyl n-piperazinecarboxylate, ethyl 1-piperazinecarboxylate, n-carbethoxypiperazine, 1-carbethoxypiperazine, 1-carboethoxypiperazine, 1-piperazinecarboxylic acid, ethyl ester, n-ethoxycarbonyl piperazine, ethylcarbonyl piperazine, 1-ethoxycarbonyl piperazine, ethyl 1-piperazinocarboxylate |
PubChem CID | 8431 |
IUPAC Name | ethyl piperazine-1-carboxylate |
SMILES | CCOC(=O)N1CCNCC1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10205110
|
Thermo Scientific Acros
118870250 |
25 g |
£28.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10316510
|
Thermo Scientific
118871000 |
100 g |
£85.90
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10463045
|
Thermo Scientific Acros
118872500 |
250 g |
£202.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
120-43-4 | |
158.2 | |
LNOQURRKNJKKBU-UHFFFAOYSA-N | |
8431 | |
CCOC(=O)N1CCNCC1 |
C7H14N2O2 | |
MFCD00005964 | |
ethyl n-piperazinecarboxylate, ethyl 1-piperazinecarboxylate, n-carbethoxypiperazine, 1-carbethoxypiperazine, 1-carboethoxypiperazine, 1-piperazinecarboxylic acid, ethyl ester, n-ethoxycarbonyl piperazine, ethylcarbonyl piperazine, 1-ethoxycarbonyl piperazine, ethyl 1-piperazinocarboxylate | |
ethyl piperazine-1-carboxylate |
Specifications
120-43-4 | |
1.0800g/mL | |
>110°C | |
98.5% min. (GC) | |
C7H14N2O2 | |
MFCD00005964 | |
1.08 | |
Solubility in water: freely soluble | |
CCOC(=O)N1CCNCC1 | |
158.2 | |
158.2 | |
Oily Liquid |
Yellow | |
273°C | |
Authentic | |
Glass Bottle | |
1.4760 to 1.478 | |
23, II, 9 | |
ethyl n-piperazinecarboxylate, ethyl 1-piperazinecarboxylate, n-carbethoxypiperazine, 1-carbethoxypiperazine, 1-carboethoxypiperazine, 1-piperazinecarboxylic acid, ethyl ester, n-ethoxycarbonyl piperazine, ethylcarbonyl piperazine, 1-ethoxycarbonyl piperazine, ethyl 1-piperazinocarboxylate | |
LNOQURRKNJKKBU-UHFFFAOYSA-N | |
ethyl piperazine-1-carboxylate | |
8431 | |
99% | |
Ethyl N-piperazinecarboxylate |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 204-395-2
- Ethyl N-piperazinecarboxylate
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only