missing translation for 'onlineSavingsMsg'
Learn More

Ethyl methanesulfonate, 99%

Product Code. 10398050 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
10g
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10398050 25 g Glass bottle 25g
10521461 10 g Glass bottle 10g
10145643 100 g Glass bottle 100g
11934561 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10398050 Supplier Thermo Scientific Acros Supplier No. 205260250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 62-50-0 | C3H8O3S | 124.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62-50-0
Molecular Formula C3H8O3S
Molecular Weight (g/mol) 124.15
MDL Number MFCD00007559
InChI Key PLUBXMRUUVWRLT-UHFFFAOYSA-N
Synonym ethyl mesylate, ethyl methanesulphonate, half-myleran, methanesulfonic acid, ethyl ester, methanesulfonic acid ethyl ester, methylsulfonic acid, ethyl ester, rcra waste number u119, ethyl methansulphonate, ethyl ester of methylsulfonic acid, ethyl ester of methanesulfonic acid
PubChem CID 6113
ChEBI CHEBI:23994
IUPAC Name ethyl methanesulfonate
SMILES CCOS(=O)(=O)C

Specifications

Density 1.1600g/mL
Boiling Point 213.0°C to 214.0°C
Flash Point 100°C
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3SO3C2H5
Refractive Index 1.4170 to 1.4190
Quantity 25 g
Beilstein 04,5
Merck Index 15,3882
Specific Gravity 1.16
Solubility Information Solubility in water: 50-100g/L (27°C)
Formula Weight 124.15
Percent Purity 99%
Chemical Name or Material Ethyl methanesulfonate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethyl methanesulfonate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 1A
  • Germ cell mutagenicity Category 1A
Hazard Statement
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.